Assay Details
| Gene Symbol: MMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082280.1 | NP_001075749.1 | 9 - 10 | 1386 | 107 | |
| GenBank mRNA | M25664.1 | - | 9 - 10 | 1386 | 107 | |
More Information
Set Membership: |
|

MMP3
Genomic Map