Assay Details
| Gene Symbol: FMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082284.1 | NP_001075753.1 | 7 - 8 | 1264 | 63 | |
| XM_008263973.2 | XP_008262195.2 | 7 - 8 | 1308 | 63 | ||
| XM_017345488.1 | XP_017200977.1 | 3 - 4 | 738 | 63 | ||
| GenBank mRNA | M32029.1 | - | 7 - 8 | 1264 | 63 | |
More Information
Set Membership: |
|

FMO2
Genomic Map