Assay Details
| Gene Symbol: WARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082288.1 | NP_001075757.1 | 979 | 93 | ||
| XM_008251449.2 | XP_008249671.1 | 983 | 93 | |||
| GenBank mRNA | M33460.1 | - | 979 | 93 | ||
More Information
Set Membership: |
|

WARS1
Genomic Map