Assay Details
| Gene Symbol: CAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082305.1 | NP_001075774.1 | 2 - 3 | 305 | 71 | |
| XM_008260452.2 | XP_008258674.1 | 5 - 6 | 919 | 71 | ||
| XM_017343635.1 | XP_017199124.1 | 4 - 5 | 338 | 71 | ||
| XM_017343636.1 | XP_017199125.1 | 4 - 5 | 977 | 71 | ||
| GenBank mRNA | M73998.1 | - | 2 - 3 | 305 | 71 | |
More Information
Set Membership: |
|

CAMP
Genomic Map