Assay Details
| Gene Symbol: CAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082305.1 | NP_001075774.1 | 3 - 4 | 385 | 65 | |
| XM_008260452.2 | XP_008258674.1 | 6 - 7 | 999 | 65 | ||
| XM_017343635.1 | XP_017199124.1 | 5 - 6 | 418 | 65 | ||
| XM_017343636.1 | XP_017199125.1 | 5 - 6 | 1057 | 65 | ||
| GenBank mRNA | M73998.1 | - | 3 - 4 | 385 | 65 | |
More Information
Set Membership: |
|

CAMP
Genomic Map