Assay Details
| Gene Symbol: SFTPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082322.1 | NP_001075791.1 | 64 | 152 | ||
| XM_008249865.2 | XP_008248087.2 | 108 | 152 | |||
| GenBank mRNA | S51597.1 | - | 64 | 152 | ||
| S51983.1 | - | 64 | 152 | |||
| X65078.1 | - | 50 | 152 | |||
More Information
Set Membership: |
|

SFTPC
Genomic Map