Assay Details
| Gene Symbol: LGALS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082338.1 | NP_001075807.1 | 3 - 4 | 422 | 81 | |
| GenBank mRNA | U06470.1 | - | 3 - 4 | 422 | 81 | |
More Information
Set Membership: |
|

LGALS3
Genomic Map