Assay Details
| Gene Symbol: CA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082372.1 | NP_001075841.1 | 2 - 3 | 309 | 108 | |
| GenBank mRNA | L48928.1 | - | 2 - 3 | 268 | 108 | |
| U58871.1 | - | 2 - 3 | 309 | 108 | ||
More Information
Set Membership: |
|

CA4
Genomic Map