Assay Details
| Gene Symbol: IP6K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082377.1 | NP_001075846.1 | 4 - 5 | 770 | 73 | |
| GenBank mRNA | U74297.1 | - | 4 - 5 | 770 | 73 | |
More Information
Set Membership: |
|

IP6K2
Genomic Map