Assay Details
| Gene Symbol: CCKAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082383.1 | NP_001075852.1 | 1 - 2 | 166 | 81 | |
| GenBank mRNA | U86601.1 | - | 1 - 2 | 166 | 81 | |
More Information
Set Membership: |
|

CCKAR
Genomic Map