Assay Details
| Gene Symbol: PTGS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082388.1 | NP_001075857.1 | 8 - 9 | 1313 | 107 | |
| GenBank mRNA | U97696.1 | - | 8 - 9 | 1313 | 107 | |
More Information
Set Membership: |
|

PTGS2
Genomic Map