Assay Details
| Gene Symbol: CNGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082394.1 | NP_001075863.1 | 467 | 76 | ||
| XM_017350856.1 | XP_017206345.1 | 478 | 76 | |||
| GenBank mRNA | X59668.1 | - | 467 | 76 | ||
More Information
Set Membership: |
|

CNGA2
Genomic Map