Assay Details
| Gene Symbol: TP53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082404.1 | NP_001075873.1 | 7 - 8 | 894 | 72 | |
| XM_008270660.2 | XP_008268882.1 | 7 - 8 | 888 | 72 | ||
| XM_017348809.1 | XP_017204298.1 | 7 - 8 | 947 | 72 | ||
| GenBank mRNA | X90592.1 | - | 894 | 72 | ||
More Information
Set Membership: |
|

TP53
Genomic Map