Assay Details
| Gene Symbol: CAPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082644.1 | NP_001076113.1 | 135 | 113 | ||
| GenBank mRNA | X72874.1 | - | 135 | 113 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAPS| Gene Symbol: CAPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082644.1 | NP_001076113.1 | 135 | 113 | ||
| GenBank mRNA | X72874.1 | - | 135 | 113 | ||
Set Membership: |
|