Assay Details
| Gene Symbol: CST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082706.1 | NP_001076175.1 | 2 - 3 | 399 | 77 | |
| GenBank mRNA | AB009342.1 | - | 2 - 3 | 399 | 77 | |
More Information
Set Membership: |
|

CST3
Genomic Map