Assay Details
| Gene Symbol: PPP1R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082744.2 | NP_001076213.1 | 3 - 4 | 577 | 70 | |
| XM_017346649.1 | XP_017202138.1 | 3 - 4 | 483 | 70 | ||
| GenBank mRNA | L20858.1 | - | 3 - 4 | 575 | 70 | |
| M95581.1 | - | 3 - 4 | 312 | 70 | ||
More Information
Set Membership: |
|

PPP1R2
Genomic Map