Assay Details
| Gene Symbol: PLAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082769.1 | NP_001076238.1 | 1482 | 56 | ||
| XM_017350327.1 | XP_017205816.1 | 1487 | 56 | |||
| XM_017350328.1 | XP_017205817.1 | 1402 | 56 | |||
| GenBank mRNA | AY029518.1 | - | 1482 | 56 | ||
More Information
Set Membership: |
|

PLAT
Genomic Map