Assay Details
| Gene Symbol: MMP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082771.1 | NP_001076240.1 | 5 - 6 | 805 | 68 | |
| GenBank mRNA | AB006779.1 | - | 5 - 6 | 790 | 68 | |
| U88652.1 | - | 5 - 6 | 805 | 68 | ||
More Information
Set Membership: |
|

MMP12
Genomic Map