Assay Details
| Gene Symbol: YBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082785.1 | NP_001076254.1 | 1 | 1108 | 105 | |
| GenBank mRNA | U16821.1 | - | 1 | 1127 | 105 | |
| Gene Symbol: YBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346398.1 | XP_017201887.1 | 1 | 990 | 105 | |
More Information
Set Membership: |
|

YBX1
Genomic Map