Assay Details
| Gene Symbol: MMP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082793.1 | NP_001076262.1 | 4 - 5 | 691 | 57 | |
| XM_017347901.1 | XP_017203390.1 | 4 - 5 | 963 | 57 | ||
| GenBank mRNA | U83918.1 | - | 4 - 5 | 691 | 57 | |
More Information
Set Membership: |
|

MMP14
Genomic Map