Assay Details
| Gene Symbol: ETS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082661.1 | NP_001076130.1 | 3 - 4 | 520 | 67 | |
| GenBank mRNA | AY561276.1 | - | 3 - 4 | 520 | 67 | |
More Information
Set Membership: |
|

ETS1
Genomic Map