Assay Details
| Gene Symbol: PRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082675.1 | NP_001076144.1 | 3 - 4 | 362 | 57 | |
| XM_017344779.1 | XP_017200268.1 | 3 - 4 | 423 | 57 | ||
| GenBank mRNA | U27199.1 | - | 3 - 4 | 362 | 57 | |
More Information
Set Membership: |
|

PRL
Genomic Map