Assay Details
| Gene Symbol: SULT1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082711.1 | NP_001076180.1 | 4 - 5 | 429 | 69 | |
| GenBank mRNA | AF026304.1 | - | 4 - 5 | 429 | 69 | |
More Information
Set Membership: |
|

SULT1C2
Genomic Map