Assay Details
| Gene Symbol: DNASE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082740.1 | NP_001076209.1 | 405 | 56 | ||
| GenBank mRNA | D82875.1 | - | 405 | 56 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNASE1| Gene Symbol: DNASE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082740.1 | NP_001076209.1 | 405 | 56 | ||
| GenBank mRNA | D82875.1 | - | 405 | 56 | ||
Set Membership: |
|