Assay Details
| Gene Symbol: MGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082745.1 | NP_001076214.1 | 1 | 1154 | 75 | |
| GenBank mRNA | M57301.1 | - | 1 | 1154 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGAT1| Gene Symbol: MGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082745.1 | NP_001076214.1 | 1 | 1154 | 75 | |
| GenBank mRNA | M57301.1 | - | 1 | 1154 | 75 | |
Set Membership: |
|