Assay Details
| Gene Symbol: POU5F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099957.1 | NP_001093427.1 | 1 - 2 | 450 | 89 | |
| GenBank mRNA | EF062856.1 | - | 1 - 2 | 450 | 89 | |
More Information
Set Membership: |
|

POU5F1
Genomic Map