Assay Details
| Gene Symbol: CRYGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099960.1 | NP_001093430.1 | 2 - 3 | 311 | 153 | |
| GenBank mRNA | EF457885.1 | - | 2 - 3 | 311 | 153 | |
More Information
Set Membership: |
|

CRYGS
Genomic Map