Assay Details
| Gene Symbol: ADH1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101704.1 | NP_001095174.1 | 3 - 4 | 294 | 120 | |
| GenBank mRNA | X69799.1 | - | 294 | 120 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADH1A| Gene Symbol: ADH1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101704.1 | NP_001095174.1 | 3 - 4 | 294 | 120 | |
| GenBank mRNA | X69799.1 | - | 294 | 120 | ||
Set Membership: |
|