Assay Details
| Gene Symbol: PPP2CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101707.1 | NP_001095177.1 | 6 - 7 | 857 | 71 | |
| GenBank mRNA | Y00763.1 | - | 857 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP2CB| Gene Symbol: PPP2CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101707.1 | NP_001095177.1 | 6 - 7 | 857 | 71 | |
| GenBank mRNA | Y00763.1 | - | 857 | 71 | ||
Set Membership: |
|