Assay Details
| Gene Symbol: PRKCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101724.1 | NP_001095194.1 | 14 - 15 | 1809 | 58 | |
| GenBank mRNA | X04796.1 | - | 1809 | 58 | ||
More Information
Set Membership: |
|

PRKCA
Genomic Map