Assay Details
| Gene Symbol: LOC100125995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109819.1 | NP_001103289.1 | 664 | 69 | ||
| XM_008250334.2 | XP_008248556.1 | 663 | 69 | |||
| XM_008250335.2 | XP_008248557.1 | 663 | 69 | |||
| XM_017338587.1 | XP_017194076.1 | 663 | 69 | |||
| XM_017338589.1 | XP_017194078.1 | 663 | 69 | |||
| GenBank mRNA | EF120627.1 | - | 664 | 69 | ||
More Information
Set Membership: |
|

LOC100125995
Genomic Map