Assay Details
| Gene Symbol: PTHLH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082653.2 | NP_001076122.2 | 3 | 542 | 114 | |
| NM_001361603.1 | NP_001348532.1 | 3 | 609 | 114 | ||
| XM_008259340.2 | XP_008257562.1 | 3 | 747 | 114 | ||
| GenBank mRNA | AF300703.1 | - | 2 | 451 | 114 | |
More Information
Set Membership: |
|

PTHLH
Genomic Map