Assay Details
| Gene Symbol: CCT6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082039.1 | NP_001075508.1 | 1 | 146 | 110 | |
| GenBank mRNA | AF068483.1 | - | 1 | 146 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCT6A| Gene Symbol: CCT6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082039.1 | NP_001075508.1 | 1 | 146 | 110 | |
| GenBank mRNA | AF068483.1 | - | 1 | 146 | 110 | |
Set Membership: |
|