Assay Details
| Gene Symbol: INMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082043.1 | NP_001075512.1 | 3 | 1647 | 99 | |
| GenBank mRNA | AF077826.1 | - | 3 | 1647 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

INMT| Gene Symbol: INMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082043.1 | NP_001075512.1 | 3 | 1647 | 99 | |
| GenBank mRNA | AF077826.1 | - | 3 | 1647 | 99 | |
Set Membership: |
|