Assay Details
| Gene Symbol: BCL2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082135.1 | NP_001075604.1 | 1 | 52 | 92 | |
| XM_008256137.2 | XP_008254359.1 | 2 | 1110 | 92 | ||
| XM_008256138.2 | XP_008254360.1 | 2 | 1092 | 92 | ||
| XM_008256139.2 | XP_008254361.1 | 2 | 370 | 92 | ||
| XM_008256140.2 | XP_008254362.1 | 2 | 352 | 92 | ||
| XM_008256141.2 | XP_008254363.1 | 2 | 159 | 92 | ||
| GenBank mRNA | AY005131.1 | - | 1 | 52 | 92 | |
More Information
Set Membership: |
|

BCL2L1
Genomic Map