Assay Details
| Gene Symbol: NAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082186.1 | NP_001075655.1 | 2 | 450 | 103 | |
| GenBank mRNA | D10111.1 | - | 2 | 450 | 103 | |
| Gene Symbol: NAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171149.1 | NP_001164620.1 | 1 | 383 | 103 | |
| XM_008267255.2 | XP_008265477.1 | 3 | 563 | 103 | ||
More Information
Set Membership: |
|

NAT2
Genomic Map