Assay Details
| Gene Symbol: UGT2B14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082240.1 | NP_001075709.1 | 5 | 1246 | 94 | |
| GenBank mRNA | L01082.1 | - | 5 | 1263 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UGT2B14| Gene Symbol: UGT2B14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082240.1 | NP_001075709.1 | 5 | 1246 | 94 | |
| GenBank mRNA | L01082.1 | - | 5 | 1263 | 94 | |
Set Membership: |
|