Assay Details
| Gene Symbol: AGXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082309.1 | NP_001075778.1 | 346 | 103 | ||
| GenBank mRNA | M84647.1 | - | 346 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGXT| Gene Symbol: AGXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082309.1 | NP_001075778.1 | 346 | 103 | ||
| GenBank mRNA | M84647.1 | - | 346 | 103 | ||
Set Membership: |
|