Assay Details
| Gene Symbol: BDKRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082347.1 | NP_001075816.1 | 445 | 84 | ||
| XM_008249375.2 | XP_008247597.1 | 612 | 84 | |||
| GenBank mRNA | U20507.1 | - | 445 | 84 | ||
More Information
Set Membership: |
|

BDKRB1
Genomic Map