Assay Details
| Gene Symbol: SCN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082359.1 | NP_001075828.1 | 348 | 80 | ||
| GenBank mRNA | U35382.1 | - | 348 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCN1B| Gene Symbol: SCN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082359.1 | NP_001075828.1 | 348 | 80 | ||
| GenBank mRNA | U35382.1 | - | 348 | 80 | ||
Set Membership: |
|