Assay Details
| Gene Symbol: KCNH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082384.1 | NP_001075853.1 | 15 | 3654 | 93 | |
| GenBank mRNA | U87513.1 | - | 15 | 3654 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNH2| Gene Symbol: KCNH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082384.1 | NP_001075853.1 | 15 | 3654 | 93 | |
| GenBank mRNA | U87513.1 | - | 15 | 3654 | 93 | |
Set Membership: |
|