Assay Details
| Gene Symbol: SLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082387.1 | NP_001075856.1 | 2 | 451 | 91 | |
| XM_017340753.1 | XP_017196242.1 | 2 | 433 | 91 | ||
| GenBank mRNA | U96091.1 | - | 2 | 451 | 91 | |
More Information
Set Membership: |
|

SLN
Genomic Map