Assay Details
| Gene Symbol: CSN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082625.3 | NP_001076094.1 | 4 | 519 | 101 | |
| GenBank mRNA | Z18243.1 | - | 519 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSN3| Gene Symbol: CSN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082625.3 | NP_001076094.1 | 4 | 519 | 101 | |
| GenBank mRNA | Z18243.1 | - | 519 | 101 | ||
Set Membership: |
|