Assay Details
| Gene Symbol: ADRA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082693.1 | NP_001076162.1 | 1 | 850 | 69 | |
| GenBank mRNA | AJ512644.1 | - | 1 | 850 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADRA2A| Gene Symbol: ADRA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082693.1 | NP_001076162.1 | 1 | 850 | 69 | |
| GenBank mRNA | AJ512644.1 | - | 1 | 850 | 69 | |
Set Membership: |
|