Assay Details
Gene Symbol: LOC100009483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082725.1 | NP_001076194.1 | 2 | 149 | 87 | |
XM_017342436.1 | XP_017197925.1 | 2 | 147 | 87 | ||
GenBank mRNA | AB029494.1 | - | 2 | 96 | 87 | |
AF360872.1 | - | 2 | 149 | 87 |
Gene Symbol: ST1A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160287.1 | NP_001153759.1 | 2 | 96 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001082725.1, NM_001160287.1, XM_017342436.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
