Assay Details
| Gene Symbol: SSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082753.1 | NP_001076222.1 | 8 | 1051 | 105 | |
| GenBank mRNA | U34847.1 | - | 8 | 1051 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSR1| Gene Symbol: SSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082753.1 | NP_001076222.1 | 8 | 1051 | 105 | |
| GenBank mRNA | U34847.1 | - | 8 | 1051 | 105 | |
Set Membership: |
|