Assay Details
| Gene Symbol: PODXL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082766.1 | NP_001076235.1 | 8 | 2954 | 114 | |
| GenBank mRNA | U35239.1 | - | 8 | 2954 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PODXL| Gene Symbol: PODXL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082766.1 | NP_001076235.1 | 8 | 2954 | 114 | |
| GenBank mRNA | U35239.1 | - | 8 | 2954 | 114 | |
Set Membership: |
|