Assay Details
| Gene Symbol: NPY6R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082773.1 | NP_001076242.1 | 1 | 657 | 108 | |
| XM_008254865.1 | XP_008253087.1 | 2 | 1036 | 108 | ||
| XM_017340985.1 | XP_017196474.1 | 2 | 1225 | 108 | ||
| GenBank mRNA | D86521.1 | - | 1 | 657 | 108 | |
More Information
Set Membership: |
|

NPY6R
Genomic Map