Assay Details
| Gene Symbol: RSC1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082774.1 | NP_001076243.1 | 748 | 85 | ||
| GenBank mRNA | X82876.1 | - | 748 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RSC1A1| Gene Symbol: RSC1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082774.1 | NP_001076243.1 | 748 | 85 | ||
| GenBank mRNA | X82876.1 | - | 748 | 85 | ||
Set Membership: |
|