Assay Details
| Gene Symbol: LIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099962.1 | NP_001093432.1 | 347 | 93 | ||
| GenBank mRNA | EF457888.1 | - | 347 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIM2| Gene Symbol: LIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099962.1 | NP_001093432.1 | 347 | 93 | ||
| GenBank mRNA | EF457888.1 | - | 347 | 93 | ||
Set Membership: |
|